Order Entry
Slovenia
Orders LinkContactUsLinkComponent
 

Hydroxy-PEG10-t-butyl ester

New Product
Supplier:   Apollo Scientific
Hydroxy-PEG10-t-butyl ester
Supplier:  Apollo Scientific
New Product
Certificates
Hydroxy-PEG10-t-butyl ester

4 Options Available Below

Product Details & Documents
Synonyms:  tert-Butyl 1-hydroxy-3,6,9,12,15,18,21,24,27,30-decaoxatritriacontan-33-oate, Hydroxy-PEG10-Boc
  • CAS Number:778596-26-2
  • Formula:C₂₇H₅₄O₁₃
  • MDL Number:MFCD28385467
  • Molecular Weight:586.71 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 30,40 €
    30,40 €
    1 * 100 mg
       30,40 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 59,40 €
    59,40 €
    1 * 250 mg
       59,40 €

    Specifications:
    New Product
    New Product
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 176,00 €
    176,00 €
    1 * 1 g
       176,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 446,00 €
    446,00 €
    1 * 5 g
       446,00 €

    Specifications:
    New Product
    New Product