
α-Biotin-omega-maleinimido poly(ethylene glycol)
Catalog # APOSBIP1049-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:GHS07
Storage Temperature:Ambient
MDL Number:MFCD00282623