Order Entry
Slovenia
ContactUsLinkComponent
Sulphosuccinimido biotin
Irritant
Sulphosuccinimido biotin
Catalog # APOSBIB102-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Sulphosuccinimido biotin
Catalog # APOSBIB102-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :443.428 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:194041-65-1
Formula:C₁₄H₁₈N₃O₈S₂·Na
MDL Number:MFCD00078535