
1,2,3,3-Tetramethyl-3H-indolium iodide ≥98%, Sigma-Aldrich®
Catalog # 328979-25G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:301.17 g/mol
Melting Pt:258 °C (dec., lit.)
:GHS07
Storage Temperature:Ambient
CAS Number:5418-63-3
Formula:C₁₂H₁₆IN
MDL Number:MFCD00013333