Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Dimethylpyridinium iodide
1,4-Dimethylpyridinium iodide 99%, Sigma-Aldrich®
1,4-Dimethylpyridinium iodide 99%, Sigma-Aldrich®
1,4-Dimethylpyridinium iodide 99%, Sigma-Aldrich®
376434-25G
:
SIGMA ALDRICH MICROSCOPY
:
1,4-Dimethylpyridinium iodide 99%, Sigma-Aldrich®
376434-25G
:
SIGMA ALDRICH MICROSCOPY
:
Pk:
25 g
Seq:
3308
:
:
235.07 g/mol
:
151…154 °C (lit.)
:
GHS07
:
Ambient
:
2301-80-6
:
C₇HIN
:
MFCD00031899