Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Diazodiphenylamine sulphate
4-Diazodiphenylamine sulphate, Sigma-Aldrich®
4-Diazodiphenylamine sulphate, Sigma-Aldrich®
4-Diazodiphenylamine sulphate, Sigma-Aldrich®
201464-25G
:
SIGMA ALDRICH MICROSCOPY
:
4-Diazodiphenylamine sulphate, Sigma-Aldrich®
201464-25G
:
SIGMA ALDRICH MICROSCOPY
:
Pk:
25 g
Seq:
3308
:
:
293.30 g/mol
:
Ambient
:
4477-28-5
:
C₁₂H₉N₃·H₂SO₄
:
MFCD00011895