Leuco malachite green, powder, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
Synonyms: 4,4'-Bis(dimethylamino)triphenylmethane, 4,4'-(Phenylmethylene)bis(N,N-dimethylaniline), 4,4'-Tetramethyldiaminotriphenylmethane, 4,4'-Benzylidenebis(N,N-dimethylaniline), Leucomalachite green
CAS Number:129-73-7
:
Formula:C₂₃H₂₆N₂
MDL Number:MFCD00008315
Molecular Weight:330.48 g/mol
Melting Pt:100...102 °C (lit.)
:M-Clarity™ quality level = MQ100
EINECS:204-961-9
:GHS07
:GHS08
:GHS09
Storage Temperature:Room Temperature
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance (Colour); White to Tan, Light Blue or Light Green:Conforms to Requirements
- Appearance (Form):Powder
- Infrared spectrum:Conforms to Structure
- Wavelength, c = 0.01 g/L in CHCl₃:259 - 265 nm
- Extinction Coefficient:Conforms to Structure
- Carbon:Typically 83.0 - 84.0%
- Nitrogen:Typically 8.0 - 9.0%
Specification Test Results
Frequently Bought Together