Order Entry
Slovenia
ContactUsLinkComponent
1-Azidoadamantane 97%, Sigma-Aldrich®
Flammable
1-Azidoadamantane 97%, Sigma-Aldrich®
Certificates
undefined
Flammable
1-Azidoadamantane 97%, Sigma-Aldrich®

Specification Test Results

  • Appearance (Colour):
    White to Off-White
  • Appearance (Form):
    Crystal to Powder
  • Infrared spectrum:
    Conforms to Structure
  • Carbon:
    65.4 - 70.1%
  • Nitrogen:
    22.8 - 24.5%
  • Purity (GC):
    ≥96.5%

Specification Test Results

Frequently Bought Together