Solvent black 5, Sigma-Aldrich®
: 2-Amino-N,N-dipropylacetamide hydrochloride, Nigrosin B, alcohol soluble, Nigrosin soluble in spirit
:11099-03-9
:
:C₈H₁₉ClN₂O
:MFCD00071316
:194.703 g/mol
- Environmentally Preferable:
- λmax:565 nm
Frequently Bought Together