

Carbol fuchsin solution, Sigma-Aldrich®
Catalog # HT8018-250ML
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:250 mL
- Seq:2590
Specifications
About this item
:
:351.88 g/mol
:GHS08
:GHS07
Storage Temperature:Ambient
:1
CAS Number:4197-24-4
Formula:C₂₁H₂₂ClN₃
MDL Number:MFCD00013129