Reactive Red 120, Sigma-Aldrich®
Catalog # R0378-50G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:50 g
- Seq:3368
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance (Colour):Red to Brown
- Appearance (Form):Powder
- Solubility (Colour):Red
- Solubility (Form), 1 mg/ml, 50% EtOH:Solution
- Carbon:≥18.0 %
- Nitrogen:≥6.7 %
- Sodium (Na):≥8 %
- Dye Content:≥50 %
- Recommended Retest Period:6 Years
Specification Test Results
About this item
:
:1469.98 g/mol
:M-Clarity™ quality level = MQ200
EINECS:263-351-0
:GHS08
Storage Temperature:Room Temperature
CAS Number:61951-82-4
Formula:C₄₄H₂₄Cl₂N₁₄Na₆O₂S₆
MDL Number:MFCD00132157