Order Entry
Slovenia
ContactUsLinkComponent
Biotin ethylenediamine hydrobromide ≥95% (by TLC), Sigma-Aldrich®
Biotin ethylenediamine hydrobromide ≥95% (by TLC), Sigma-Aldrich®
  B9181-25MG
 :  
undefined
Biotin ethylenediamine hydrobromide ≥95% (by TLC), Sigma-Aldrich®
  B9181-25MG
 :  

 

  • Pk:
    25 mg
  • Seq:
    2315

 

 

 :
 :367.31 g/mol
 :Refrigerator
 :216299-38-6
 :C₁₂H₂₃BrN₄O₂S
 :MFCD00467807