Order Entry
Slovenia
ContactUsLinkComponent
 
bisbenzimide H-33342 trihydrochloride (Hoechst 33342) ≥98% (by HPLC, TLC), Sigma-Aldrich®
 
undefined
bisbenzimide H-33342 trihydrochloride (Hoechst 33342) ≥98% (by HPLC, TLC), Sigma-Aldrich®
 

 

  • Environmentally Preferable:

 

 

  • Appearance (Form):
    Powder
  • Appearance (Colour):
    Yellow
  • Solubility (Solvent):
    Water
  • Solubility (Conc):
    49.00 - 51.00 mg/ml
  • Solubility (Turbidity):
    Clear to Very Slightly Hazy
  • Solubility (Colour):
    Yellow to Brown
  • NMR (Solvent):
    D2O
  • Identity by NMR:
    Consistent
  • Purity (TLC):
    ≥98.00%
  • Purity (HPLC):
    ≥98.00%

 

 

Frequently Bought Together