Order Entry
Slovenia
ContactUsLinkComponent
6-(2,2-Dicyanovinyl)-N-(2-hydroxyethyl)-1,2,3,4-tetrahydroquinoline, Sigma-Aldrich®
Irritant
6-(2,2-Dicyanovinyl)-N-(2-hydroxyethyl)-1,2,3,4-tetrahydroquinoline, Sigma-Aldrich®
Catalog # D5928-5MG
CAS Number:  
undefined
Irritant
6-(2,2-Dicyanovinyl)-N-(2-hydroxyethyl)-1,2,3,4-tetrahydroquinoline, Sigma-Aldrich®
Catalog # D5928-5MG
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :253.3 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:142978-25-4
Formula:C₁₅H₁₅N₃O
MDL Number:MFCD00274418