Sleeve protectors, A4 cleanroom, KIMTECH PURE®
Catalog # KIMB49815_CM
Supplier: KIMBERLY CLARK
Specifications
- Garment application:Cleanroom
- Colour:White
- Length:533 mm
- Environmentally Preferable:Waste Reducing
- Pk:200
Specifications
About this item
The KIMTECH PURE® A4 Sleeve Protectors helps keep workers and the workplace safe through advanced barrier protection that shields the person and the garment from hazardous chemicals and biologics.
- Thumb loops for full wrist coverage
- Reinforced seams
- Blue indicator line and tag to facilitate donning and doffing
- Bulk packaged for convenience and waste reduction
- Recyclable through the RightCycle™
Ideal for situations where full coverage isn't required, but full protection is advanced barrier fabric provides enhanced protection against liquid exposure.
These reduces the risk of exposure and contamination by providing an additional layer of protection between gloves and lab coat sleeves.