Order Entry
Slovenia
Orders LinkContactUsLinkComponent
VWR® Labtop Coolers
VWR® Labtop Coolers
VWR® Labtop Coolers
Supplier:  VWR Collection
Certificates
VWR® Labtop Coolers
VWR® Labtop Coolers
3 Options Available Below
Product Details & Documents
−20 °C mini labtop cooler PC, non toxic gel: The −20 °C mini labtop cooler holds 12×0,5 or 1,5 ml microcentrifuge tubes at below −20 °C for over one hour. Ideal for bench top use or transport. Filled with non toxic gel. Prior to use, place in −25 °C freezer for at least 24 hours, store at −25 °C between use.
−20 °C maxi labtop cooler PC, non toxic gel
The −20 °C maxi labtop cooler, holds 32×0,5 or 1,5 ml microcentrifuge tubes at below −20 °C for over 1 hour. Suitable for bench top use or transport. Wire handle secures top while carrying. Cooler and lid filled with non toxic gel. Store at −25 °C between use.
−20 °C labtop cooler PC, non toxic gel
The −20 °C labtop cooler maintains temperatures between −20 and −15 °C for up to 4 hours. Holds 20× 1,5 or 2,0 ml microcentrifuge tubes or cryovials. Inserts included hold 0,2 to 0,5 ml microcentrifuge tubes. Printed grid, keyed lid, locking handle and non skid feet. Store at −25 °C between use.
The −20 °C labtop cooler maintains temperatures between −20 and −15 °C for up to four hours. Polycarbonate. Holds twenty 1,5 to 2,0 ml microcentrifuge tubes or cryovials. Inserts included hold 0,2 to 0,5 ml microcentrifuge tubes. Printed grid, keyed lid, locking handle, non skid feet. Store at −25 °C between use.
Product Family Options
Product Information
  • No. of Holes
    Dimensions
    Availability
    Price
  • 479-0642
    −20 °C mini labtop cooler
    −20 °C mini labtop cooler
    12
    151×108×125 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 146,00 €
    146,00 €
    1 * 1 items
       146,00 €
  • 479-0643
    −20 °C labtop cooler
    −20 °C labtop cooler
    20
    151×108×125 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 169,00 €
    169,00 €
    1 * 1 items
       169,00 €
  • 479-0641
    −20 °C maxi labtop cooler
    −20 °C maxi labtop cooler
    32
    243×157×146 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 181,00 €
    181,00 €
    1 * 1 items
       181,00 €