Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Rain jackets, S571
Rain jackets, S571
Supplier:   Portwest
Rain jackets, S571
Supplier:  Portwest
Certificates
Rain jackets, S571
4 Options Available Below
Product Details & Documents
A classic jacket with good fit to streamline and reduce the bulkiness women often feel in a fourseason jacket. The longer line back panel and discreet waist adjustment make this a contemporary silhouette and give a flattering smooth front jacket.
  • 3 in 1 waterproof and windproof jacket
  • Extremely water resistant fabric finish, water beads away from fabric surface
  • Taped seams to provide additional protection
  • Interactive inner garment
  • Interactive fleece which can be worn as a separate jacket for multi use
  • Five pockets for ample storage which are zipped phone pocket
  • Two-way zip for quick and easy access
  • Hook and loop closure for easy access
The following Rainwear collections offer an outstanding choice of designs and colours, suitable for many end users. The best quality fabrics and high technology construction techniques are used throughout, guaranteeing the ultimate in all-weather protection. Tested to withstand the rigours of everyday wear, each garment has been designed and manufactured with care to ensure total satisfaction wear after wear.
100% polyester, Pongee PVC coated shell fabric, 100% polyester lining fabric with 100% polyester fleece removable fabric.
Certifications: EN 343 Class 3:1 (WP 5,000 mm).
Specifications for Product Family
Specifications
  • Catalog No:
  • PRTWS571NARXS
  • PRTWS571NARS
  • PRTWS571NARL
  • PRTWS571NARXL
  • Model:
  • S571
  • S571
  • S571
  • S571
  • Garment application:
  • General Purpose
  • General Purpose
  • General Purpose
  • General Purpose
  • Risk area:
  • Rain (waterproof)
  • Rain (waterproof)
  • Rain (waterproof)
  • Rain (waterproof)
  • Gender:
  • For ladies
  • For ladies
  • For ladies
  • For ladies
  • Sleeve or leg length:
  • Long
  • Long
  • Long
  • Long
  • Closure type:
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Fabric:
  • 100% polyester
  • 100% polyester
  • 100% polyester
  • 100% polyester
  • Weight:
  • 550 g
  • 550 g
  • 550 g
  • 550 g
Specifications
Product Family Options
Product Information
  • Colour
    Clothing Size
    Availability
    Price
  • Dark blue
    XS
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 113,00 €
    113,00 €
    1 * 1 items
       113,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    PRTWS571NARXS
    Model
    S571
    Garment application
    General Purpose
    Risk area
    Rain (waterproof)
    Gender
    For ladies
    Sleeve or leg length
    Long
    Closure type
    Zip closure
    Fabric
    100% polyester
    Weight
    550 g
  • Dark blue
    S
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 113,00 €
    113,00 €
    1 * 1 items
       113,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    PRTWS571NARS
    Model
    S571
    Garment application
    General Purpose
    Risk area
    Rain (waterproof)
    Gender
    For ladies
    Sleeve or leg length
    Long
    Closure type
    Zip closure
    Fabric
    100% polyester
    Weight
    550 g
  • Dark blue
    L
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 113,00 €
    113,00 €
    1 * 1 items
       113,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    PRTWS571NARL
    Model
    S571
    Garment application
    General Purpose
    Risk area
    Rain (waterproof)
    Gender
    For ladies
    Sleeve or leg length
    Long
    Closure type
    Zip closure
    Fabric
    100% polyester
    Weight
    550 g
  • Dark blue
    XL
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 113,00 €
    113,00 €
    1 * 1 items
       113,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    PRTWS571NARXL
    Model
    S571
    Garment application
    General Purpose
    Risk area
    Rain (waterproof)
    Gender
    For ladies
    Sleeve or leg length
    Long
    Closure type
    Zip closure
    Fabric
    100% polyester
    Weight
    550 g