
Bismuth chloride oxide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7787-59-9
:
Formula:BiClO
MDL Number:MFCD00010895
:260.43 g/mol
Density:7.72 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:232-122-7
:GHS07
Frequently Bought Together