Silicon dioxide (Silica, vitreous), acid-washed, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:60676-86-0
:
Formula:SiO₂
MDL Number:MFCD00011232
:60.08 g/mol
Boiling Pt:2230 °C (1013 hPa)
Melting Pt:1710 °C
Density:2.66 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:231-545-4
Frequently Bought Together