Aluminium carbide, Sigma-Aldrich®
SIAL241873-100G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
:
:143.96 g/mol
:12,00345
:2100 °C
:2.36 g/cm³ (20 °C)
:215-076-2
:1394
:4.3,II
:M-Clarity™ quality level = MQ100
:1299-86-1
:C₃Al₄
:MFCD00003425