

Aluminium carbide ≥99% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1299-86-1
:
Formula:C₃Al₄
MDL Number:MFCD00003425
:143.96 g/mol
Merck Index:14,335
Melting Pt:2100 °C
Density:2.36
Storage Temperature:Ambient
EINECS:215-076-2
UN:1394
ADR:4.3,II
:GHS02
:GHS07
Specification Test Results
- Total Metal Impurities:0.9% max
Specification Test Results
Frequently Bought Together