

Potassium iodate
Catalog # PROONCSGC76209
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:214 g/mol
Merck Index:13,07726
Melting Pt:560 °C
Density:3.98 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-831-9
UN:1479
ADR:5.1,II
:GHS03
:GHS07
CAS Number:7758-05-6
Formula:KlO₃
MDL Number:MFCD00011406