Order Entry
Slovenia
ContactUsLinkComponent
PIPES monosodium salt
PIPES monosodium salt
Catalog # ROTH9153.1
Supplier:  Roth Carl
CAS Number:  
PIPES monosodium salt
Catalog # ROTH9153.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :324.35 g/mol
Storage Temperature:Ambient
EINECS:233-005-3
CAS Number:10010-67-0
Formula:C₈H₁₇N₂NaO₆S₂
MDL Number:MFCD00065472