4'-Ethylacetophenone, Sigma-Aldrich®
Catalog # SIAL226750-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:148.21
Boiling Pt:240 °C (1013 hPa)
Density:0.9719 g/cm³ (0 °C)
:M-Clarity™ quality level = MQ100
EINECS:213-326-5
CAS Number:937-30-4
Formula:C₁₀H₁₂O
MDL Number:MFCD00009262