Histodenz™, Sigma-Aldrich®
Catalog # D2158-100G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
About this item
Synonyms: Iohexol, N1,N3-Bis(2,3-dihydroxypropyl)-5-(N-(2,3-dihydroxypropyl)acetamido)-2,4,6-triiodoisophthalamide, 5-[Acetyl(2,3-dihydroxypropyl)amino]-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide, N,N'-Bis(2,3-dihydroxypropyl)-5-[N-(2,3-dihydroxypropyl)acetamido]-2,4,6-triiodoisophthalamide
:
:821.142 g/mol
Storage Temperature:Ambient
CAS Number:66108-95-0
Formula:C₁₉H₂₆I₃N₃O₉
MDL Number:MFCD00077732