Ficoll® solution 20% in H2O, Sigma-Aldrich®
:26873-85-8
:
:MFCD00081599
:1.055…1.075 g/mL
:M-Clarity™ quality level = MQ200
:Refrigerator
- Appearance (Turbidity):Clear
- Appearance (Colour):Colorless
- Appearance (Form):Liquid
- Density:1.055 - 1.075 g/ml
- Registered Trademark:Ficoll is a registered trademark of GE Healthcare.
Frequently Bought Together