Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Rinse solution no. 2
Rinse solution no. 2, Sigma-Aldrich®
Rinse solution no. 2, Sigma-Aldrich®
Flammable
Rinse solution no. 2, Sigma-Aldrich®
Catalog # RS2-900ML
Supplier:
SIGMA ALDRICH MICROSCOPY
CAS Number:
Expand image
Flammable
Rinse solution no. 2, Sigma-Aldrich®
Catalog # RS2-900ML
Supplier:
SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
Pk:
900 mL
Seq:
2815
Specifications
About this item
Product Details
Documentation
:
:
GHS02
Storage Temperature:
Ambient
:
1
MDL Number:
MFCD00011332
Product Details
Documentation