Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
L-Lysine
L-Lysine
L-Lysine
L-Lysine
555-0029
:
Cytiva (Formerly Pall Lab)
:
L-Lysine
555-0029
:
Cytiva (Formerly Pall Lab)
:
Pack type:
Glass bottle
Pk:
1 L
Seq:
5037
:
:
146.19 g/mol
:
13,05656
:
290 °C (1013 hPa)
:
196 °C
:
200-294-2
:
56-87-1
:
C₆H₁₄N₂O₂
:
MFCD00064433