Order Entry
Slovenia
ContactUsLinkComponent
Europium triacetate hydrate 99%
Europium triacetate hydrate 99%
Catalog # A18069.03
CAS Number:  
Europium triacetate hydrate 99%
Catalog # A18069.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

 :
 :347.11 g/mol
Storage Temperature:Ambient
EINECS:214-670-9
CAS Number:62667-64-5
Formula:C₆H₉EuO₆·H₂O
MDL Number:MFCD00150115