

Hydrochloric acid ≥36.5% ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Merck Index:14,4780
Boiling Pt:81.5…110 °C
Melting Pt:–30 °C
Density:1.19
Storage Temperature:Ambient
EINECS:231-595-7
ADR:8,II
:36.46 g/mol
UN:1789
:GHS05
:GHS07
Related Information
Frequently Bought Together