Bromothymol blue 0.04% in ethanol, TECHNICAL
Catalog # 34303.234
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 mL
- Seq:2590
Specifications
Specification Test Results
- Identification:Passes test
Specification Test Results
About this item
:
ADR:3,II
:624.39 g/mol
EINECS:200-971-2
UN:1170
:GHS02
Storage Temperature:Ambient
CAS Number:76-59-5
Formula:C₂₇H₂₈Br₂O₅S
MDL Number:MFCD00005872