Order Entry
Slovenia
ContactUsLinkComponent
VWR® Soap Solution According to Boutron-Boudet
VWR® Soap Solution According to Boutron-Boudet
Catalog # 31700.293
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® Soap Solution According to Boutron-Boudet
Catalog # 31700.293
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Description:
    Soap solution according to Boutron-Boudet
Specifications
About this item
Solution for water hardness testing.
0,104 ml corresponds to 1 hydrotimetric degree for a 40 ml water sample