



White spirit, 150…200 °C, TECHNICAL
28963.368
:
- Pack type:Plastic bottle, fluorinated
- Pk:5 L
- Seq:5190
- Refractive index (20°C):1.425 - 1.450
- Benzene:Max. 10 ppm
:
:150…200 °C (1013 hPa)
:–15 °C
:0.77 g/cm³ (20 °C)
:42 °C
:919-857-5
:1268
:3,III
:01-2119463258-33
:GHS02
:GHS08
:GHS07
:GHS09
:64742-48-9