
Dithizone ≥98.0%, AnalaR NORMAPUR® analytical reagent
Catalog # 23570.106
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Assay:≥ 98.0 %
- Appearance:Purple to dark purple or black powder
- Specific absorptivity 1% / 1cm (at 605 nm):min. 1522
- Relative absorption (A 605nm/at 445nm; Chloroform):min. 2.5
- Sulphated ash:max. 0.2%
- Heavy metals (as Pb):max. 0.001%
Specification Test Results
About this item
:
:256.33 g/mol
Merck Index:13,03413
Boiling Pt:406 °C (1013 hPa)
Melting Pt:168 °C
EINECS:200-454-1
:GHS07
CAS Number:60-10-6
Formula:C₆H₅NHNHCSN=NC₆H₅
MDL Number:MFCD00003025