
Norflurano, Cryo-Jet Lamb's freezing aerosol
361852T
:
- Pack type:Aerosol can
- Pk:1
- Seq:0001
- Purity % (w/w):99,5%
- Appearance:Clear and colourless liquefied gas
- Water Content ppm (w/w):≤10 ppm
- Non-condensable Gas (Vapour phase) %v/v:1,5%
- High Boiling Residue % (w/w):≤0,010 %
- Acidity (As HCl) ppm (w/w):≤1 ppm
- Chloride:No visible turbidity
:
:114.043 g/mol
:13,04734
:–19 °C
:–101 °C
:∼1.2 g/cm³ (20 °C)
:Ambient
:212-377-0
:3163
:2.2,
:GHS04
:29118-24-9
:C₃H₂F₄