VWR®, Cloud Point standard, Certified Reference Material
99104.230
:
- Pk:250 mL
- Description:Cloud Point Standard
- Environmentally Preferable:
- Temperature:–7,6 °C
Cloud Point Certified Reference Materials are for the verification of laboratory instruments used in testing petroleum and derivative products for Cloud Point according to IP 219, ISO 3015, EN 23015, ASTM D2500 and other equivalent test methodology.
- Tested in accordance with ASTM / EN or IP test method protocols
- Low level of uncertainty achieves maximum data accuracy
- Supplied in tamper-evident UN compliant security pack with 12 month shelf life
- Fully traceable to international standards
: IP 219, ISO 3015, EN 23015, ASTM D2500
Fully cerified in accordance with UKAS and ISO/IEC 17034.
Fully cerified in accordance with UKAS and ISO/IEC 17034.
: The temperature value given in the ordering table below is the nominal temperature for the cloud point value - actual batch values are given in the certification documents supplied with the product and also on the label.