Industrial gloves, ABERDEEN, TRIUS®
Supplier: TRIUS SAFETY
Certificates
About this item
Impact and oil-resistant, cut and sewn synthetic gloves offer protection against shock, cuts, and fat. The ABERDEEN series solves multiple problems while ensuring high visibility.
- High cut resistance
- Impact protection
- Palm and thumb reinforcement
- Oil-proof
- High visibility
ABERDEEN gloves are designed for use in wet and oily environments, with risks of cut or impact. This protective glove features a palm made of a 3-layer complex (synthetic leather/cut protection/nitrile coating textile), a back made of cut-resistant and oilrepellent high visibility textile with protection against shock, and a highly breathable Hyprene® wrist. They are washable and durable.
Certifications: EN 388:2016 (2X44EP)
Packaging: Bundle of 5 pairs.
Specifications
- Catalog No:
- TRIUABERDEEN/9
- TRIUABERDEEN/8
- TRIUABERDEEN/11
- TRIUABERDEEN/10
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Liner material:
- Nitrile/nylon
- Nitrile/nylon
- Nitrile/nylon
- Nitrile/nylon
- Finish:
- Impact protection on back
- Impact protection on back
- Impact protection on back
- Impact protection on back
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Cuff length:
- 7 cm
- 7 cm
- 7 cm
- 7 cm
- Cuff thickness:
- 15 mil
- 15 mil
- 15 mil
- 15 mil
- Finger thickness:
- 16 mil
- 16 mil
- 16 mil
- 16 mil
- Palm thickness:
- 16 mil
- 16 mil
- 16 mil
- 16 mil
- EN norm:
- EN 388:2016 (2X44EP)
- EN 388:2016 (2X44EP)
- EN 388:2016 (2X44EP)
- EN 388:2016 (2X44EP)