Order Entry
Slovenia
ContactUsLinkComponent
Lithium bis(oxalato)borate, Sigma-Aldrich®
Irritant
Lithium bis(oxalato)borate, Sigma-Aldrich®
Catalog # SIAL757136-25G
Supplier:  Merck
CAS Number:  
Irritant
Lithium bis(oxalato)borate, Sigma-Aldrich®
Catalog # SIAL757136-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance (Color):
    White
  • Appearance (Form):
    Powder or Crystals
  • X-Ray Diffraction :
    Conforms to Structure

Specification Test Results

About this item

 :
 :193.79 g/mol
 :GHS07
CAS Number:244761-29-3
Formula:C₄BLiO₈
MDL Number:MFCD07776904