Order Entry
Slovenia
ContactUsLinkComponent
Allyl(chloro)[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(II)
Allyl(chloro)[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(II)
Catalog # 46747.03
CAS Number:  
Allyl(chloro)[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(II)
Catalog # 46747.03
CAS Number:  

Specifications

  • Pk:
    1 g

Specifications

About this item

 :
 :893.666 g/mol
Storage Temperature:Ambient
CAS Number:1248656-98-5
Formula:C₅₁H₅₂ClFePPd
MDL Number:MFCD25372546