Organic reference standards, Serotonin, analytical standard, Supelco®
Catalog # SIAL14927-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- CAS No.:50-67-9
- Description:Serotonin
- Formula:C₁₀H₁₂N₂O
- Storage temperature:2…8 °C
- Description:Serotonin