ESD safety shoes, Velcro® fastening, uvex 2 xenova® S1 Trainer, 9504
Supplier: UVEX
Certificates
About this item
Metal-free, lightweight, improved fit and optimum cushioning, padding significantly increase comfort, made of micro velour upper, soft padded collar and outsole made from dual-density PUR sole with excellent slip resistance. Sole materials are free from silicones, plasticisers and other paint-wetting impairment substances.
- Climazone technology for improved comfort
- Double individually adjustable velcro
- Breathable textile lining
- Removable anti-static comfort insole with moisture control system; with a moisture-wicking system
- Anti-slip sole with shock absorption at heel and front of the foot
- 100% metal-free xenova® toecap, compact design, anatomical shape, good lateral stability, no thermal conductivity
- Fulfils ESD requirements: resistance to ground inferior to 35 megaohms
- Suitable for indoor and outdoor works
Contains uvex xenova® plastic cap which is the safety feature of this shoes. Suitable for people allergic to chrome, made from synthetic materials.
Colour: Black/red
Certifications: EN 61340 (ESD); EN ISO 20345:2011; S1, SRC
Frequently Bought Together