Order Entry
Slovenia
ContactUsLinkComponent
Rat tail collagen coating solution
Rat tail collagen coating solution
Catalog # SIAL122-20
Supplier:  Merck
CAS Number:  
Rat tail collagen coating solution
Catalog # SIAL122-20
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    Rat tail collagen coating solution, 20 ml

Specifications

About this item

Rat tail collagen coating solution enhances cell attachment and proliferation, or promotion of cell-specific morphology and function. Commonly used for endothelial cells, hepatocytes, muscle cells, and a variety of other cell types. This collagen solution is prepared in 0,02 M acetic acid at 50 ug/ml and sterile filtered.

Coat tissue cultureware with rat tail collagen solution to culture surface ratio of 5 ug/cm². For example: 125 μg (2,5 ml) for a T-25 flask; 10,5 μg (210 μl) for each well of a 24-well plate.

Coated tissue cultureware may be used immediately or air-dried and stored at 4 °C for up to one week.