VACUUBRAND VARIO® technology - frees you up for other tasks. The VACUU·SELECT controller simplifies your work and allows VARIO select chemistry pumping systems to deliver unparalleled precision. The pumps run whisper quiet and respond to demand in real time by matching pumping speed to process conditions. The intuitive VACUU·SELECT controller includes predefined applications for all common vacuum processes.
- VACUU·SELECT vacuum controller, with graphical user interface and pre-defined applications, simplifies work in the laboratory
- Automatic boiling point detection and adjustment of the vacuum for short process times
- Extraordinary diaphragm life for minimum operational and servicing costs
- Excellent environmental compatibility through low energy consumption and efficient solvent recovery
The inlet separator, made of glass with a robust protective coating, prevents particles and liquid droplets from entering the pump, preventing damage and extending pump life. The exhaust vapour condenser provides nearly 100% recovery of solvents for economical recycling and environmental protection.
PC 3016 VARIO select
These powerful pumps feature exceptionally high pumping speed. The PC 3016 VARIO select pumping unit is a proven solution for evaporations with large amounts of low boiling point solvents.
PC 3012 VARIO select
With its combination of high pumping speed and ultimate vacuum, this pump is suitable for demanding applications, even with high boiling point solvents. This pump is regularly used to support floor-standing rotary evaporators, vacuum drying ovens, distillation columns, and other similar work in miniplants, kilo labs and scale-up facilities.
PC 3010 VARIO select
With its combination of high pumping speed and fractional millibar ultimate vacuum, this pump is suitable for demanding applications, even with high boiling point solvents at low temperatures. This pump is regularly used to support floor-standing rotary evaporators, vacuum drying ovens, distillation columns, and other similar work in miniplants, kilo labs and scale-up facilities.