Order Entry
Slovenia
ContactUsLinkComponent
Flame-retardant shirts, long sleeves, 8850
Flame-retardant shirts, long sleeves, 8850
Supplier:  UVEX
Certificates
undefined
Flame-retardant shirts, long sleeves, 8850
Supplier:  UVEX
Specifications
  • Catalog No:
  • UVEX8968742
  • UVEX8968744
  • UVEX8968738
  • UVEX8968740
  • UVEX8968748
  • UVEX8968746
  • Model:
  • 8850
  • 8850
  • 8850
  • 8850
  • 8850
  • 8850
  • Garment application:
  • Fire protection
  • Fire protection
  • Fire protection
  • Fire protection
  • Fire protection
  • Fire protection
  • Risk area:
  • Flame-retardant
  • Flame-retardant
  • Flame-retardant
  • Flame-retardant
  • Flame-retardant
  • Flame-retardant
  • Gender:
  • For men
  • For men
  • For men
  • For men
  • For men
  • For men
  • Sleeve or leg length:
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Closure type:
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Fabric:
  • 49% modacryl, 49% viscose and 2% carbon fibres
  • 49% modacryl, 49% viscose and 2% carbon fibres
  • 49% modacryl, 49% viscose and 2% carbon fibres
  • 49% modacryl, 49% viscose and 2% carbon fibres
  • 49% modacryl, 49% viscose and 2% carbon fibres
  • 49% modacryl, 49% viscose and 2% carbon fibres
  • Weight:
  • 165 g/m²
  • 165 g/m²
  • 165 g/m²
  • 165 g/m²
  • 165 g/m²
  • 165 g/m²