Ethyl cellulose 48.0 - 49.5% (ethoxyl content)
Supplier: Merck
Certificates
About this item
CAS Number:9004-57-3
:
MDL Number:MFCD00131037
Density:1.14 g/ml (25 °C) (lit.)
Storage Temperature:Ambient
Specification Test Results
- Assay:48.0 - 49.5% (w/w) ethoxyl basis
- Form:Powder
- Refractive Index:n20/D 1.47 (lit.)
Specification Test Results
Frequently Bought Together