Calibration standard, ProteoMass™ LTQ/FT-hybrid ESI Pos. mode cal mix
SIALMSCAL5-1EA
: Merck
:
- Pk:1
- Standards:Calibration standard
- Description:ProteoMass™ LTQ/FT-hybrid ESI Pos. mode cal mix
- M-Clarity quality level:MQ100
- Pk Info:1× 10 ml
This mixture can be used as a calibration standard for the mass spectrometric analysis of serum samples of patients affected by mugwort pollen allergy induced by pollen extract of Artemisia vulgaris.
- Ready-to-use
- Developed and qualified specifically for use with the LTQ based Thermo Scientific hybrid instruments with ESI source and Thermo Scientific Exactive
- Conveniently packaged
- Store at –20 °C