Order Entry
Slovenia
ContactUsLinkComponent
Dehydrated culture media, WL nutrient broth
Dehydrated culture media, WL nutrient broth
Catalog # SIALW2261-500G
Supplier:  Merck
CAS Number:  
Dehydrated culture media, WL nutrient broth
Catalog # SIALW2261-500G
Supplier:  Merck
CAS Number:  
Specifications
  • Pk:
    500 g
  • Standards:
    -
  • Description:
    WL nutrient broth
Specifications
About this item
WL nutrient broth consists of casein enzymatic hydrolysate (5,00 g/l), yeast extract (4,00 g/l), dextrose (50,00 g/l), monopotassium phosphate (0.55 g/l), potassium chloride (0.425 g/l), calcium chloride (0.125 g/l), manganese sulphate (0.125 g/l), ferric chloride (0.0025 g/l), manganese sulphate (0.0025 g/l) and bromocresol green (0.022 g/l).
Used for the cultivation of bacteria encountered in breweries and industrial fermentations.