Order Entry
Slovenia
ContactUsLinkComponent
Poly-L-lactic acid (PLA), granules 3 mm (nominal) biopolymer
Poly-L-lactic acid (PLA), granules 3 mm (nominal) biopolymer
Catalog # SIALGF45989881-1EA
Supplier:  Merck
CAS Number:  
Poly-L-lactic acid (PLA), granules 3 mm (nominal) biopolymer
Catalog # SIALGF45989881-1EA
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
CAS Number:26100-51-6
Formula:(C₃H₆O₃)n
MDL Number:MFCD00081867