Order Entry
Slovenia
ContactUsLinkComponent
 
Cysteinyl leukotriene express ELISA kit
 
undefined
Cysteinyl leukotriene express ELISA kit

 

  • Developmental time:
    60 - 90 min
  • Mid point:
    70 - 110 pg/ml (50% B/B₀)
  • Detection range:
    7,8 - 1000 pg/ml
  • Storage temperature:
    −80 °C
  • Sensitivity:
    20 pg/ml (80% B/B₀)
  • Incubation time:
    2 hr

 

Frequently Bought Together