8021/502.2 surrogate mix #1
Catalog # REST30463
Supplier: Restek
CAS Number:
Specifications
- Concentration:2,000 µg/ml each in P and T methanol
- Pk:1
- Description:8021/502.2 Surrogate Mix #1
- Pk Info:1 ml/ampoule
Specifications
About this item
The mix consists of 1-bromo-2-chloroethane, 1-chloro-3-fluorobenzene and fluorobenzene.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.